Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Stanniocalcin 1/STC-1 Rabbit anti-Human, DyLight 680, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297046FR
Description
Stanniocalcin 1/STC-1 Polyclonal specifically detects Stanniocalcin 1/STC-1 in Human samples. It is validated for ELISA.Specifications
Stanniocalcin 1/STC-1 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
DyLight 680 | |
stanniocalcin 1, stanniocalcin-1, STCSTC-1 | |
Produced in rabbits immunized with purified, recombinant Stanniocalcin 1/STC-1 (NP_003146.1; Met1-Ala247). | |
0.1 mL | |
Signal Transduction | |
6781 | |
Store at 4°C in the dark. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title