Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stannous Chloride, Dihydrate, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
2.5kg
500g
Packaging:
Poly Bottle

Chemical Identifiers

10025-69-1
SnCl2·2H2O

High quality chemicals for laboratory and specialized industrial use

    Suitable for Mercury Determination
CAS 10025-69-1
Molecular Formula SnCl2·2H2O
Formula Weight 225.63
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 2.71kg
Quantity 500g
Chemical Name or Material Stannous Chloride, Dihydrate
Grade Baker Analyzed™ ACS Reagent, Meets ACS reagent chemical requirements