Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stannous Chloride, Dihydrate, Crystal, Reagent, ACS, 98-103%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
6 x 125 g
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
10025-69-1
Cl2H4O2Sn
225.64
FWPIDFUJEMBDLS-UHFFFAOYSA-L
λ²-tin(2+) dihydrate dichloride
O.O.[Cl-].[Cl-].[Sn++]
10025-69-1
Cl2H4O2Sn
225.64
FWPIDFUJEMBDLS-UHFFFAOYSA-L
λ²-tin(2+) dihydrate dichloride
O.O.[Cl-].[Cl-].[Sn++]

S1535, 10025-69-1, SnCl2•2H2O

Spectrum™ Stannous Chloride, Dihydrate, Crystal, Reagent, ACS is also known as Tin Chloride, has a white solid crystal like aspect. Mostly used in electrolytic baths destined for tin-plating, it is also widely used as a reducing agent. In the textile industry it is employed as a mordant to increase the brightness of certain colors in the fabric. It is also employed by metal enthusiasts to determine platinum and gold group metals. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.

CAS Max % 1
Percent Purity 98 to 103%
Calcium (Ca) 5e-005
Iron (Fe) 3e-005
Lead (Pb) 0.0001
Packaging Poly Bottle
Potassium (K) 5e-005
Sodium (Na) 0.0001
Quantity 6 x 125 g