Learn More
MAPA™ StanSolv™ A-18 Unlined Nitrile Gloves
Ultra-long chemical protection
Manufacturer:  MAPA™ 480439
 View more versions of this product
Catalog No. 11-392-5A
Add to cart
Description
Description
  • Z-Patter grip channels liquids away from glove surface for an enhanced grip in wet applications
  • Nitrile compound provides resistance to snags, punctures, abrasions and cuts
  • Chlorinated finish provides easy on/off
  • Straight cuff
Specifications
Specifications
Safety
Flat
Nitrile
22 mil
Green
45.7 cm
Embossed
handling drums of chemical products, treating/degreasing metals with aggressive solvents, pesticide application, maintenance, timber treatment and finishing
9
Powder-free
18 in.