Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPA™ StanSolv™ A-18 Unlined Nitrile Gloves

Ultra-long 18 in. heavy weight nitrile gloves feature a unique Z-pattern grip for a secure grip, and are double dipped for a leak-proof barrier against chemical permeation.

Manufacturer:  MAPA™ 480438

 View more versions of this product

Catalog No. 19-042-652


Add to cart
This item is not returnable. View return policy

Description

Description

  • Z-Pattern grip channels liquids away from glove surface for an enhanced grip in wet applications
  • Nitrile compound provides resistance to snags, punctures, abrasions and cuts
  • Chlorinated finish provides easy on/off
  • Straight cuff

Specifications

Specifications

Green
Flat
Nitrile
StanSolv™ A-18
18 in.
Cat III CE 0334
EN 388:4102X
Type A
Bacteria, Fungi only
22 mil
Yes
Handling drums of chemical products, treating/degreasing metals with aggressive solvents, pesticide application, maintenance, timber treatment and finishing
Unlined
480438
8
45.7 cm
21 CFR 170-199
No
AJKOPT
Yes
0.55 mm
Z-pattern
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit