Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPA™ StanSolv™ A-18 Unlined Nitrile Gloves

Ultra-long chemical protection

$254.00

Specifications

Length (English) 18 in.
Length (Metric) 45.7 cm
Thickness (English) 22 mil
Coating Material Embossed
Color Green
View More Specs

Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-042-652
View Documents
MAPA™
480438
8 Dozen of 12 for $254.00
Add to cart
 
11-392-5A
View Documents
MAPA™
480439
9 Dozen of 12 for $254.00
Add to cart
 
11-392-5B
View Documents
MAPA™
480430
10 Dozen of 12 for $254.00
Add to cart
 
11-392-5C
View Documents
MAPA™
480431
11 Dozen of 12 for $254.00
Add to cart
 
Description

Description

  • Z-Patter grip channels liquids away from glove surface for an enhanced grip in wet applications
  • Nitrile compound provides resistance to snags, punctures, abrasions and cuts
  • Chlorinated finish provides easy on/off
  • Straight cuff

Specifications

Specifications

18 in.
22 mil
Green
Flat
45.7 cm
Embossed
Nitrile
Powder-free
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit