Learn More
3M™ Petrifilm™ Staph Express Count Plate, Sold by AquaPhoenix Scientific
Provides Staphylococcus aureus results
$167.00
Specifications
Detects | Staphylococcus aureus |
---|---|
Product Type | Count Plate |
For Use With (Application) | Food processors and contract labs, Facilitating enumeration of S. aureus |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
S30734
|
3M™ Petrifilm™
6490 |
50 Pack |
Each for $167.00
|
|
|||||
S30710
|
3M™ Petrifilm™
6491 |
500 Pack | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Results in 24 hours
- Plates are sample-ready, eliminating time-consuming steps of preparing media/agar dishes
- Provides accurate, efficient, consistent results for food processors and contract labs
- Unique system to facilitates enumeration of S. aureus
- S. aureus appears as distinctive red-violet colonies
- This plate has been awarded Official Methods of Analysis (OMA) #2003.07 for processed and prepared foods, #2003.8 for select dairy foods and #2003.11 for Select meat, seafood , poultry and NF VALIDATION certificates: 3M-01/9-04/03 A and 3M-01/9-04/03 B for All human foods and Pet food by AFNOR Certification
Note:
- Other colonies have to be checked using Staph express disk
Specifications
Staphylococcus aureus | |
Count Plate | |
Food processors and contract labs, Facilitating enumeration of S. aureus |