Learn More
Innovating Science LyoQuick Freeze Dried Bacteria Cultures
Easy to use lyophillized bacterial cultures for use in classroom for educational purposes. Innovating Science LyoQuick Freeze Dried Bacteria Cultures, eliminates several steps required for rehydration of freeze-dried cultures, thus reducing the risk of contamination.
Manufacturer: Innovating Science IS3606
This item is not returnable.
View return policy
Description
- Each tube contains two ampules, one containing freeze-dried bacteria and one self-contained sterile rehydration broth
- To use, pinch tube to break internal ampules and let freezedried organisms rehydrate, then dispense rehydrated organisms into broth or onto agar through dropper tip
- Available for seven different types of bacteria
Specifications
White | |
LyoQuick Freeze Dried Bacteria | |
1 | |
20 | |
13 |
Format: Freeze Dried | |
Staphylococcus epidermidis | |
4 x 0.5 x 0.5 in. | |
14 | |
7 |