Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Starch, for analysis, soluble, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
5kg
Packaging:
Plastic bottle
Plastic drum
9005-84-9
C12H22O11
342.297
MFCD00082026
GUBGYTABKSRVRQ-ASMJPISFSA-N
alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose
439341
CHEBI:18167
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O
9005-84-9
C12H22O11
342.297
MFCD00082026
GUBGYTABKSRVRQ-ASMJPISFSA-N
alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose
439341
CHEBI:18167
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O
Merck Index 15, 8926
Identification positive
Physical Form Amorphous Powder
Infrared Spectrum Authentic
Grade Analysis
Loss on Drying 10% max.
Packaging Plastic drum
Sensitivity passes
Solubility Solubility in water: 50g/L (90°C).
Sulfated Ash 0.5% max.
Color White
Melting Point 256.0°C to 258.0°C
pH 5 to 7.5 (2% soln. at 25°C)
Quantity 5kg
Chemical Name or Material Starch, Soluble, For analysis
Show More Show Less