Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Starch, Hydrolyzed, Ultrapure, Affymetrix/USB™

Click to view available options
Quantity:
100g
1kg
Packaging:
Plastic bottle
9005-25-8
C12H22O11
342.297
MFCD00082026
GUBGYTABKSRVRQ-ASMJPISFSA-N
alpha-maltose,maltose,starch, soluble,glcalpha1-4glca,unii-15sug9ad26,glcalpha1-4glcalpha,amylodextrin,starch solution,alpha-malt sugar,4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose
439341
CHEBI:18167
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O
9005-25-8
C12H22O11
342.297
MFCD00082026
GUBGYTABKSRVRQ-ASMJPISFSA-N
alpha-maltose,maltose,starch, soluble,glcalpha1-4glca,unii-15sug9ad26,glcalpha1-4glcalpha,amylodextrin,starch solution,alpha-malt sugar,4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose
439341
CHEBI:18167
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O
Physical Form Powder
Packaging Plastic bottle
Quantity 100g
Chemical Name or Material Starch