Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Starch Indicator, Aqueous Solution, 0.5% (w/v), Ricca Chemical

Click to view available options
Quantity:
1L
Packaging:
Poly natural
9005-84-9
C12H22O11
342.297
GUBGYTABKSRVRQ-ASMJPISFSA-N
439341
CHEBI:18167
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O
9005-84-9
C12H22O11
342.297
GUBGYTABKSRVRQ-ASMJPISFSA-N
439341
CHEBI:18167
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O
Appearance Translucent liquid
Assay Percent Range 0.5% (w/v)
Physical Form Liquid
Percent Purity 0.50%
Odor Slight potato-like odor
Packaging Poly natural
pH Range 5 to 7 (2% solution)
Solubility Solubility in Water: Infinite
Quantity 1L

Recommended Storage : General