Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Starch Indicator, Preserved with Salicylic Acid (Mercury Free), Certified, 0.5%, LabChem™

Supplier:  LabChem, Inc. LC253002

 View more versions of this product

Catalog No. LC253002


Add to Cart

Description

Description

Specifications

Specifications

Starch Indicator
99.4, 0.5, 0.1
Soluble in water
0.5%
Poly Bottle
Passes Test
1g/mL
Liquid
Preserved with Salicylic Acid (Mercury Free)
9005-25-8, 69-72-7
Passes Test
Certified
1g/mL
Fume; Carbon monoxide; Carbon dioxide
1 L
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit