Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Starch indicator solution 1%, Acculute Standard Volumetric Solution, Final volume 1L, Alfa Aesar™

For Iodometric Titrations

Manufacturer:  Alfa Aesar 88632LS

Quantity 6 units

 View more versions of this product

Catalog No. AA88632LS


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

9005-84-9,7732-18-5
100%
Starch indicator solution
Liquid
MFCD00082026

Specifications

9005-84-9,7732-18-5
100%
Starch: 20%; Water: 80%
6 units
Liquid
MFCD00082026
17mm Hg at 20°C
Starch indicator solution
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit