Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Starch Iodide Paste TS, (U.S.P. Test Solution), Spectrum™

Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass Bottle

S-219, 9005-25-8

Spectrum™ Starch Iodide Paste TS, (U.S.P. Test Solution) is a suspension of reagents, prepared to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays. This product is manufactured by Spectrum.

CAS 9005-84-8,7681-11-0,7646-85-7
CAS Max % 0.001,0.001,0.998
Grade Chemical Solution
Packaging Amber Glass Bottle
Quantity 500 mL