Learn More
BioVision IncSupplier Diversity Partner Starch Colorimetric/Fluorometric Assay Kit: Starch Assay Kit: Colorimetric & Fluorometric Assay to measure Starch levels in a variety of Samples within 40 min. Simple & Convenient. Detection Range: 0.0004 to 2 mg/ml. 100 Assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K647100
Starch is a complex carbohydrate consisting of a large number of glucose units. All plants contain starch, present as amylose, (linear α-1,4 linked polymer) and amylopectin, (highly α-1,6 branched α-1,4 polymer). Starch generally contains 0-25% amylose and 75–100% amylopectin. The Starch Assay Kit provides an easy, accurate assay to measure starch levels in a variety of samples. In the assay, starch is hydrolyzed to glucose which is oxidized to generate color (570 nm) and fluorescence (Ex/Em = 535/587 nm). The assay can detect starch at 0.0004 to 2 mg/ml. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9816