Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ STARD10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA536947
Description
This antibody detects endogenous protein at a molecular weight of 35 kDa. Purity is >95% by SDS-PAGE.
STARD10 gene ontology annotations related to this gene include bile acid secretion; positive regulation of peroxisome proliferator activated receptor signaling pathway.
Specifications
STARD10 | |
Polyclonal | |
Unconjugated | |
STARD10 | |
Antigen NY-CO-28; AV048538; CGI52; CGI-52; NY-C0-28; NY-CO-28; PCTP2; PC-TP2; Pctpl; PCTP-L; PCTP-like protein; Sdccag28; Sdccagg28; serologically defined colon cancer antigen 10; serologically defined colon cancer antigen 28; Serologically defined colon cancer antigen 28 homolog; StAR related lipid transfer domain containing 10; STARD10; StAR-related lipid transfer (START) domain containing 10; StAR-related lipid transfer domain containing 10; stAR-related lipid transfer protein 10; START domain containing 10; START domain-containing protein 10; TISP-81 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10809, 293150, 56018 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9JMD3, Q9Y365 | |
STARD10 | |
Synthetic peptide corresponding to amino acids 105-150 of human STARD10. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction