Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STARD10, Rabbit anti-Human,Mouse,Rat, Janelia Fluor 646, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP278818JF646
Description
STARD10 Polyclonal antibody specifically detects STARD10 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).Specifications
STARD10 | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Antigen NY-CO-28, CGI-52, MGC14401, NY-CO-28, PCTP2, PCTP-L, PCTP-like protein, Serologically defined colon cancer antigen 28SDCCAG28, StARD10, StAR-related lipid transfer (START) domain containing 10, StAR-related lipid transfer protein 10, START domain containing 10, START domain-containing protein 10 | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Affinity Purified, Conjugated | |
STARD10 | |
Full length recombinant human STARD10 protein (amino acids 1-291) [UniProt Q9Y365] | |
0.1 mL | |
Store at 4°C in the dark. | |
Polyclonal |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title