Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STARD8 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP169097
Description
STARD8 Polyclonal specifically detects STARD8 in Human samples. It is validated for Western Blot.Specifications
STARD8 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
ARHGAP38, Deleted in liver cancer 3 protein, DKFZp686H1668, DLC3DLC-3, KIAA0189stAR-related lipid transfer protein 8, StARD8, StAR-related lipid transfer (START) domain containing 8, START domain containing 8, START domain-containing protein 8, STARTGAP3, START-GAP3 | |
Rabbit | |
122 kDa | |
100 μL | |
Primary | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
STARD8 | |
Synthetic peptides corresponding to STARD8 (StAR-related lipid transfer (START) domain containing 8) The peptide sequence was selected from the N terminal of STARD8. Peptide sequence SSDRPLLSPTQGQEGPQDKAKKRHRNRSFLKHLESLRRKEKSGSQQAEPK. | |
Affinity Purified | |
RUO | |
9754 | |
Human, Canine | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title