Learn More
Starfish
SCI_ED
Pisaster sp.
 View more versions of this product
Catalog No. S920310
Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.
Description
Description
  • Easy-to-dissect Asteroidea-starfish from the genus Pisaster is a representative invertebrate deuterosome
  • Single-injected specimens have latex in the water vascular system
  • Select vacuum packed or in pails
Specifications
Specifications
Starfish
6 to 8 in.
10 Pk.
Plain
15.2 to 20.3 cm
Pail
Product Suggestions
Product Suggestions