Learn More
Heidolph™ StarFish™ Glassware - Soxhlet
SDP
Heidolph™ StarFish™ Glassware - Soxhlets are available in various volumes, and socket sizes
$416.12 - $1172.14
Specifications
Type Soxhlet
View More Specs
Products 4
Catalog Number Mfr. No. Capacity (Metric) Size Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Size Price Quantity & Availability  
05-404-109
SDP
View Documents
Heidolph™
015890820
20 mL Socket: 24/29
Each for $416.12
Only null left
Add to Cart
05-404-110
SDP
View Documents
Heidolph™
015890830
40 mL Socket: 29/32
Each for $1,169.05
Only null left
Add to Cart
05-404-111
SDP
View Documents
Heidolph™
015890840
60 mL Socket: 34/25
Each for $1,172.14
Only null left
Add to Cart
05-404-112
SDP
View Documents
Heidolph™
015890850
100 mL Socket: 40/38
Each for $1,171.11
Only null left
Add to Cart
Description
Description
  • Used for extracting lipids or similar components from a volatile solvent
  • Available in various volumes, and socket sizes
Specifications
Specifications
Soxhlet
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.