Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stargazin Mouse anti-Human, Mouse, Rat, Clone: S245-1, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527644

Catalog No. PIMA527644


Add to cart

Description

Description

Stargazin is a member of the transmembrane AMPAR regulatory proteins (TARP) family and is involved in glutamate receptor trafficking. It has been recently demonstrated that the interaction between stargazin and AMPA receptors is critical for the correct localization of the receptors at the synapse. Phosphorylation of the stargazin protein at Thr 321 by Protein Kinase A regulates its interaction with PSD-95 and synaptic targeting of AMPA receptors.
Specifications

Specifications

Stargazin
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
O88602, Q71RJ2, Q9Y698
CACNG2, Calcium channel voltage dependent gamma subunit 2, CaV gamma 2, Ipr328, MGC123981, MGC138502, MGC138504, Voltage-dependent calcium channel gamma-2 subunit, Neuronal voltage-gated calcium channel gamma-2 subunit, Stargazin, Stg, Stargazer, TARP, TARP gamma-2, Transmembrane AMPAR regulatory protein gamma-2, AW060990, B230105C07Rik, B930041E13Rik, Wag, Waggler
Mouse
IgG2A
100 μg
-20°C
Primary
10369, 12300, 84347
Immunocytochemistry, Immunofluorescence, Western Blot
S245-1
Unconjugated
CACNG2
Liquid
Cacng2
Fusion protein amino acids 203-323 (Cytoplasmic C-terminus) of rat TARPGamma2
Protein G
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit