Starplex™ Histoplex™ Histology Containers
Specifically engineered for the storage of histology specimens
Manufacturer: Starplex Scientific H902FL
 View more versions of this product
Catalog No. 15-182-73
  Check Availability
Add to Cart
    Specifications
    Color Clear
    Graduated Yes
    Material Polypropylene
    Label Affixed
    Closure Color Yellow
    Sterile No
    Cap Type Screw cap with O-ring
    Capacity (Metric) 90mL
  • Description & Specifications
Related Products
Product Content Feedback