Starplex Scientific™ False Bottom Analyzer Tube

Designed for high-volume analyzer aspiration probes incapable of reaching the bottoms of larger sample tubes

Manufacturer: Starplex Scientific V055VO

 View more versions of this product

Catalog No. 14-905-60

  Check Availability
Add to Cart

    Specifications

    Capacity (Metric) 5mL
    Color Amber
    For Use With (Application) Designed for high-volume analyzer aspiration probes incapable of reaching the bottoms of larger sample tubes
    Material Polypropylene
    Shape Conical
    Sterile No
    Closure Type Without
    Product Type False Bottom Analyzer Vial

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit