Learn More
Starplex Scientific Histoplex™ Tissue Cassettes with Micromesh Chamber
Innovative micromesh chamber securely retains fragments, leaving ample room for additional larger specimens
Manufacturer:  Starplex Scientific TCYELLOW
 View more versions of this product
Catalog No. 22-146-422
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Slots
Clam-Shell
Acetal Polymer
45°
Yellow
Tissue/Biopsy Cassette