Starplex™ Scientific Multitrans™ Collection and Transportation Systems

Complete collection and transport systems for viruses, Chlamydia, Mycoplasma and Ureaplasma

Manufacturer: Starplex Scientific S160COM

 View more versions of this product

Catalog No. 22-146-531

  Check Availability
Add to Cart

    Specifications

    Product Type Swab
    For Use With (Equipment) Multitrans collection and transportation system
    Includes Scored plastic shafted Dacron tipped swab, transport medium with glass beads and antibiotics, and a 10mL clarified polypropylene sleak-resistant tube with skirted screw cap
    Quantity 100 Case
    Storage Requirements 2 to 25°C

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit