Learn More
Starplex™ Scientific Starswab II™ Collection and Transport Systems: Amies Charcoal Medium
Protective overskirt on cap
Manufacturer:  Starplex™ Scientific SP140X
 View more versions of this product
Catalog No. 14-905-100
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • Designed for collecting, transporting and maintaining the viability of aerobic, anaerobic and facultative microorganisms
  • Swab tips available in single, double or straight wire format
  • Applicators have color-coded caps with overskirt and biohazard plug-cap rings; extra-long swab shafts; and a labeled, round-bottom, spiral transport tube
  • Transport tubes contain 4mL of semi-solid media
  • Spiral media chamber within a seamless polypropylene tube protects sample and keeps it immersed in media during transport
  • Labels preprinted with lot number, expiration date and product code with space reserved for recording patient information
  • Gamma sterilized and wrapped in sterile, easy-to-peel envelope
Amies Charcoal Medium
  • Improves the survival of fastidious organisms (especially Neisseria gonorrhoeae) by neutralizing any toxic components present
Specifications
Specifications
Collecting, transporting and maintaining the viability of aerobic, anaerobic and facultative microorganisms
Amies Plus Gel
Bag
Plastic
Rayon
Amies Plus reduced Charcoal
Single
100 Pack
4 to 25°C
Safety and Handling
Safety and Handling
ShelfLife : 2 Year(s)