Learn More
Starplex™ Scientific Starswab II™ Collection and Transport Systems: Amies Charcoal Medium
Protective overskirt on cap
Manufacturer:  Starplex Scientific SP131D
 View more versions of this product
Catalog No. 14-905-94
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Black
Modified Amies Charcoal Gel
50 Pack
Double
Collecting, transporting and maintaining the viability of aerobic, anaerobic and facultative microorganisms
Amies Charcoal
Bag
Plastic
4 to 25°C
Rayon
Safety and Handling
ShelfLife : 2 Year(s)