Learn More
Starplex™ Scientific Starswab II™ Collection and Transport Systems: Amies Clear Medium
Protective overskirt on cap
Manufacturer:  Starplex™ Scientific SP130FW
 View more versions of this product
Catalog No. 14-905-90
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • Designed for collecting, transporting and maintaining the viability of aerobic, anaerobic and facultative microorganisms
  • Swab tips available in single, double or straight wire format
  • Applicators have color-coded caps with overskirt and biohazard plug-cap rings; extra-long swab shafts; and a labeled, round-bottom, spiral transport tube
  • Transport tubes contain 4mL of semi-solid media
  • Spiral media chamber within a seamless polypropylene tube protects sample and keeps it immersed in media during transport
  • Labels preprinted with lot number, expiration date and product code with space reserved for recording patient information
  • Gamma sterilized and wrapped in sterile, easy-to-peel envelope
Amies Clear Medium
  • General-purpose transport medium for recovery of a wide range of organisms, including certain anaerobes
  • Provides excellent survival characteristics for transport and gram stain compatibility
Specifications
Specifications
Collecting, transporting and maintaining the viability of aerobic, anaerobic and facultative microorganisms
Modified Amies Clear Gel
Orange
100 Case
4 to 25°C
Flexible wire
Amies Clear
Bag
Flex Wire
Rayon
Safety and Handling
Safety and Handling
ShelfLife : 2 Year(s)