Learn More
Starplex™ Scientific Urine Transfer Kit and Accessories
For storage and transport of urine and other liquid samples
$269.10 - $408.25
Specifications
Sterility Sterile
View More Specs
Includes: Leak resistant sterile container with needle free spout, straw, collar, castille wipe
Products 2
Catalog Number Mfr. No. For Use With (Application) Includes Product Type Specs Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Application) Includes Product Type Specs Price Quantity & Availability  
22-046-419
View Documents
Starplex™ Scientific
B1202SO
For Urine transfer kit, For storage and transport of Urine and other liquid samples Needle free spout, straw, collar, castille wipe Specimen Container
Case of 300 for $269.10
Add to cart
22-046-418
View Documents
Starplex™ Scientific
UTK1202SO
For safe collection, transport and storage of Urine Leak resistant 120mL container with needle free spout, straw, collar, castille wipe Transfer Kit
Case of 100 for $408.25
Add to cart
Description
Description
  • Kit includes 120mL sterile container and straw
  • Extra containers and straws also available
Specifications
Specifications
Sterile