Starplex™ Scientific Urine Transfer Kit and Accessories
For storage and transport of urine and other liquid samples
Manufacturer: Starplex Scientific UTK1202SO
 View more versions of this product
Catalog No. 22-046-418
  Check Availability
Add to Cart
    Specifications
    Product Type Transfer Kit
    For Use With (Application) For safe collection, transport and storage of Urine
    Sterility Yes
    Includes Leak resistant 120mL container with needle free spout, straw, collar, castille wipe
  • Description & Specifications
Related Products
Product Content Feedback