Products for Science Education Starry Night™ High School

Specifications

For Use With Application Explorations and understanding of the Universe
For Use With Equipment DVD player and TV, or computer with DVD player.
Format Individual DVD
Grade 9 to 12
Includes Teacher's Guide
View More Specs

Includes: Award-winning Starry Night planetarium software, SkyTheater DVD, and introductory astronomy book

  • Products 3
  • Description & Specifications
Products
Catalog Number Mfr. No. User License Type Price Quantity & Availability    

S96656AND

Simulation Curriculum Corp.
EDUHS-60-C1U SINGLE
Individual

S96656BND

Simulation Curriculum Corp.
EDUHS-60-C1U 5SEAT
5 seat

S96656CND

Simulation Curriculum Corp.
EDUHS-60-C1U 30SEAT
30 seat
Descriptions & Specifications

Related Products