Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT1 Mouse anti-Human, Clone: AT1FA, Abnova™

Mouse monoclonal antibody raised against recombinant human STAT1.

Manufacturer:  Abnova Corporation MAB12470

Catalog No. 89-934-970


Add to cart

Description

Description

The protein encoded by this gene is a member of the STAT protein family. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. This protein can be activated by various ligands including interferon-alpha, interferon-gamma, EGF, PDGF and IL6. This protein mediates the expression of a variety of genes, which is thought to be important for cell viability in response to different cell stimuli and pathogens. Two alternatively spliced transcript variants encoding distinct isoforms have been described. [provided by RefSeq]

Specifications

Specifications

STAT1
AT1FA
Mouse monoclonal antibody raised against recombinant human STAT1.
In PBS, pH 7.4 (10% glycerol, 0.02% sodium azide)
DKFZp686B04100/ISGF-3/STAT91
Mouse
IgG1, kappa
50 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
ELISA, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
ELISA Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Liquid
STAT1
Recombinant protein corresponding to amino acids 1-712 of human STAT1.
Protein A purification
RUO
Primary
6772
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit