Learn More
Stat1 (pY701) Mouse, BV421, Clone: 4a, BD Horizon™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 566238
Catalog No. BDB566238
Add to cart
Description
Description
Stat1 (pY701) Monoclonal antibody specifically detects Stat1 (pY701) in Human, Mouse, Rat samples. It is validated for Flow cytometry (Intracellular Staining)) .
Specifications
Specifications
Stat1 (pY701)
4a
Vol. Per test: 5μL
Purified
Murine
Mouse IgG2a
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions, and unconjugated antibody and free BD Horizon™ BV421 were removed.
Monoclonal
Flow Cytometry, Flow Cytometry (Intracellular Staining)
Brilliant Violet 421
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Signal Transducer and Activator of Transcription-1; STAT91; ISGF-3; CANDF7
Phosphorylated Human Stat1 Peptide
25 Tests
Cell Biology
Primary
Human, Murine, Rat
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.