Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT1, Rabbit anti-Human, Clone: 3E6, Abnova™

Rabbit Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB19115

Catalog No. 89-335-949


Add to cart

Specifications

Specifications

STAT1
3E6
Flow Cytometry (1 ug/mL-0.001 ug/mL ) The optimal working dilution should be determined by the end user.
signal transducer and activator of transcription 1, 91kDa
DKFZp686B04100|ISGF-3|STAT91
Rabbit
IgG1k
RUO
Antibody
Monoclonal
Human, Mouse
Flow Cytometry
Unconjugated
In PBS (50% glycerol, 0.1% BSA, 0.02% sodium azide).
Liquid
STAT1
A synthetic phosphopeptide corresponding to residues surrounding Tyr701 of human phospho STAT1.
200 μL
Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Primary
6772
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit