Learn More
Stat2 Monoclonal Antibody (19G8.H2.H6), Rockland™

Mouse Monoclonal Antibody
$174.73 - $651.91
Specifications
Antigen | Stat2 |
---|---|
Clone | 19G8.H2.H6 |
Concentration | 1 mg/mL |
Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-0581
![]() ![]() |
Rockland Immunochemicals
200301B65 |
100 μg |
Each for $651.91
|
|
|||||
50-199-0582
![]() ![]() |
Rockland Immunochemicals
200301B65S |
25 μL |
Each for $174.73
|
|
|||||
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Directed against human STAT2 protein. A BLAST analysis was used to suggest cross-reactivity with STAT2 protein from human, rat (73%) and mouse (76%), sources based on homology with the immunizing sequence. Reactivity against homologues from other sources is not known.
STAT2 is a member of the STAT protein family. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. In response to interferon (IFN), this protein forms a complex with STAT1 and IFN regulatory factor family protein p48 (ISGF3G), in which this protein acts as a transactivator, but lacks the ability to bind DNA directly. Transcription adaptor P300/CBP (EP300/CREBBP) has been shown to interact specifically with this protein, which is thought to be involved in the process of blocking IFN-alpha response by adenovirus.Specifications
Stat2 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
1600010G07Rik; AW496480; BOS_5470; IMD44; interferon alpha induced transcriptional activator; ISGF-3; MGC59816; p113; Signal transducer and activator of transcription 2; signal transducer and activator of transcription 2, 113kDa; STAT; STAT 2; STAT113; STAT2; STAT-2 | |
Stat2 | |
IgG2a κ | |
-20°C, Avoid Freeze/Thaw Cycles | |
Stat2 |
19G8.H2.H6 | |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
P52630, Q9WVL2 | |
20847, 6773 | |
This monoclonal antibody was produced by repeated immunizations with a synthetic peptide corresponding to residues near the carboxy terminus of mouse STAT2 protein. | |
Primary | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.