Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAT2 Mouse anti-Human, Clone: STAT2/2650, Novus Biologicals™
Mouse Monoclonal Antibody
$265.00
Specifications
Antigen | STAT2 |
---|---|
Clone | STAT2/2650 |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB122708
|
Novus Biologicals
NBP30761720UG |
20 μg |
Each of 1 for $265.00
|
|
|||||
Description
STAT2 Monoclonal specifically detects STAT2 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
STAT2 | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cancer, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
10 mM PBS with 0.05% BSA | |
6773 | |
IgG1 κ | |
Protein A or G purified |
STAT2/2650 | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
interferon alpha induced transcriptional activator, ISGF-3, MGC59816, p113, signal transducer and activator of transcription 2, signal transducer and activator of transcription 2, 113kD, signal transducer and activator of transcription 2, 113kDa, STAT113 | |
Recombinant full-length human STAT2 protein | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title