Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAT3 Rabbit anti-Human, Clone: SY24-08, Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer: Novus Biologicals NBP267432
Description
STAT3 Monoclonal antibody specifically detects STAT3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
STAT3 | |
SY24-08 | |
Unconjugated | |
Purified | |
Acute-phase response factor, APRFMGC16063, DNA-binding protein APRF, FLJ20882, HIES, signal transducer and activator of transcription 3, signal transducer and activator of transcription 3 (acute-phase responsefactor) | |
STAT3 | |
Recombinant Protein | |
Monoclonal | |
Protein A purified | |
Cellular Markers, Myeloid derived Suppressor Cell, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
Human,Mouse,Rat,Zebrafish |
Western Blot,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunoprecipitation,Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1:000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation, Immunohistochemistry-Paraffin 1:50-1:200 | |
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide | |
6774 | |
Rabbit | |
IgG | |
Primary | |
100μL | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title