Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT4 Mouse anti-Human, Clone: OTI2F3, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA503041

Catalog No. 50-167-4014


Add to cart

Description

Description

The STAT 4 protein is a transcription factor activated by hormone and cytokines. STAT 4 is most similar to STAT 1 (52%) and STAT 3 (47%). Functionally, STAT 4 is similar to other STAT family members in that it can be tyrosine phosphorylated by JAK 1 or JAK 2. STAT 4 forms homodimers and heterodimers with related STAT family members. Tyrosine phosphorylated STAT 4 can bind the interferon g activated site (GAS), and serine phosphorylation is also required for maximal transcriptional activity. STAT 4 expression is restricted to the thymus, spleen, and testis.
Specifications

Specifications

STAT4
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q14765
SLEB11
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6775
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI2F3
Unconjugated
STAT4
Liquid
STAT4
Full length human recombit protein of human STAT4 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit