Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAT5a/b Rabbit anti-Human, Mouse, Rat, Clone: 3J2A2, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | STAT5a/b |
---|---|
Clone | 3J2A2 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
STAT5a/b Monoclonal antibody specifically detects STAT5a/b in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
STAT5a/b | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer, Signal Transduction, Transcription Factors and Regulators | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
6776 | |
IgG | |
Affinity purified |
3J2A2 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
MGF, STAT5, STAT5A signal transducer and activator of transcription 5A | |
Recombinant fusion protein containing a sequence corresponding to amino acids 600-786 of human STAT5A (P42229). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title