Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT5A Mouse anti-Human, DyLight 405, Clone: 251610, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP266707V

Catalog No. NBP266707V


Add to cart

Description

Description

STAT5A Monoclonal antibody specifically detects STAT5A in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

STAT5A
251610
Flow Cytometry
MGF, signal transducer and activator of transcription 5A, STAT5
Mouse
IgG1
Cancer, Phospho Specific, Signal Transduction, Transcription Factors and Regulators
Primary
6776
Human
Flow Cytometry
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
STAT5A
Human STAT5a synthetic peptide, SLDSRLSPPAGLFTSARGSLS, Accession # NP_003143
0.1mL
Store at 4°C in the dark.
Monoclonal
Detects human STAT5a. This antibody does not cross-react with STAT5b.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit