Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAT5A Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$236.93 - $378.00
Specifications
Antigen | STAT5a (p Tyr694) |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10082219
|
Novus Biologicals
NB10082219 |
0.05 mL |
Each for $378.00
|
|
NB017883
|
Novus Biologicals
NB100822190.025ML |
0.025 mL |
Each for $236.93
|
|
Description
STAT5A Polyclonal specifically detects STAT5A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
STAT5a (p Tyr694) | |
Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 | |
Polyclonal | |
Rabbit | |
Phospho Specific, Signal Transduction | |
P42229 | |
6776 | |
The antiserum was produced against synthesized phosphopeptide derived from human STAT5A around the phosphorylation site of tyrosine 694 (D-G-YP-V-K). | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1.0 mg/mL | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
MGF, signal transducer and activator of transcription 5A, STAT5 | |
STAT5A | |
IgG | |
Affinity Purified | |
90 kDa |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title