Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAT5b Mouse anti-Human, Clone: STAT5B/2657, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30732320UG
Description
STAT5b Monoclonal specifically detects STAT5b in Human samples. It is validated for ELISA, Protein Array.Specifications
STAT5b | |
Monoclonal | |
0.2 mg/ml | |
ELISA 1-5 ug/ml, Protein Array | |
signal transducer and activator of transcription 5B, STAT5, transcription factor STAT5B | |
Recombinant full-length human STAT5b protein | |
20 μg | |
Phospho Specific, Signal Transduction | |
6777 | |
Store at 4°C. | |
IgG |
ELISA, Peptide Array | |
STAT5B/2657 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction