Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT5b Mouse anti-Human, Clone: STAT5B/2657, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen STAT5b
Clone STAT5B/2657
Concentration 0.2 mg/ml
Dilution ELISA 1-5 ug/ml, Protein Array
Applications ELISA, Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122120
View Documents
Novus Biologicals
NBP30732320UG
20 μg
Each for $138.37
Add to cart
 
NB122119
View Documents
Novus Biologicals
NBP30732310UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

STAT5b Monoclonal antibody specifically detects STAT5b in Human samples. It is validated for ELISA, Protein Array
Specifications

Specifications

STAT5b
0.2 mg/ml
ELISA, Peptide Array
Purified
RUO
Human
signal transducer and activator of transcription 5B, STAT5, transcription factor STAT5B
Recombinant full-length human STAT5b protein
Monoclonal
Protein A or G purified
STAT5B/2657
ELISA 1-5 ug/ml, Protein Array
Unconjugated
Mouse
Phospho Specific, Signal Transduction
10 mM PBS with 0.05% BSA
6777
IgG
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit