Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAT6 Polyclonal Antibody, Active Motif™

Rabbit Polyclonal Antibody
Supplier: Active Motif 61547

Description
STAT6 protein is a transcription factor activated by cytokines, particularly interleukin-4 and IL13. (Stat6-/-) were found to be deficient in IL-4-mediated functions including Th2 helper T-cell differentiation, expression of cell surface markers, T-cell proliferation, immunoglobulin class switching to IgE, and partial loss of IL-4 mediated proliferation. STAT6 mRNA has been detected in peripheral blood lymphocytes, colon, intestine, ovary, prostate, thymus, spleen, kidney, liver, lung and placenta. STAT6 is critically involved in Th2 immune response. STAT6 is crucial for IL-4-mediated biological responses, and naturally in two isoforms: STAT6a and STAT6c.Specifications
STAT6 | |
Polyclonal | |
Unconjugated | |
STAT6 | |
12S1644; BOS_5462; D12S1644; IL4 STAT; IL-4 Stat; IL-4-STAT; Signal transducer and activator of transcription 6; signal transducer and activator of transcription 6, interleukin-4 induced; Signal transducer and transcription activator 6; STAT; STAT, interleukin4-induced; Stat6; STAT-6; STAT6B; STAT6C; transcription factor IL-4 STAT | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6778 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Lot-Specific | |
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 | |
P42226 | |
STAT6 | |
This antibody was raised against a peptide within the C-terminal region of human STAT6. This antibody detects two isoforms of STAT6. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title