Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT6 Rabbit anti-Human, Unconjugated, Polyclonal, Active Motif™

Rabbit Polyclonal Antibody

Manufacturer:  Active MotifSupplier Diversity Partner 61548

Encompass

 View more versions of this product

Catalog No. 50-199-3485


Add to cart

Description

Description

STAT6 protein is a transcription factor activated by cytokines, particularly interleukin-4 and IL13. (Stat6-/-) were found to be deficient in IL-4-mediated functions including Th2 helper T-cell differentiation, expression of cell surface markers, T-cell proliferation, immunoglobulin class switching to IgE, and partial loss of IL-4 mediated proliferation. STAT6 mRNA has been detected in peripheral blood lymphocytes, colon, intestine, ovary, prostate, thymus, spleen, kidney, liver, lung and placenta. STAT6 is critically involved in Th2 immune response. STAT6 is crucial for IL-4-mediated biological responses, and naturally in two isoforms: STAT6a and STAT6c.
Specifications

Specifications

STAT6
Western Blot
Unconjugated
STAT6
Liquid
Rabbit
IgG
10 μL
-20°C, Avoid Freeze/Thaw Cycles
Primary
6778
STAT6
Lot-Specific
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
P42226
12S1644; BOS_5462; D12S1644; IL4 STAT; IL-4 Stat; IL-4-STAT; Signal transducer and activator of transcription 6; signal transducer and activator of transcription 6, interleukin-4 induced; Signal transducer and transcription activator 6; STAT; STAT, interleukin4-induced; Stat6; STAT-6; STAT6B; STAT6C; transcription factor IL-4 STAT
This antibody was raised against a peptide within the C-terminal region of human STAT6. This antibody detects two isoforms of STAT6.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit