Sino Biological™ STAT6 Recombinant Human Protein, His Tag Non-distribution product as customer accommodation.

Suitable for use in protein studies such as protein structure analysis and protein-protein interactions

Manufacturer: Sino Biological™ 13190H08B5

 View more versions of this product

Catalog No. 13-190-H08B5

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Structural Form Full Length, Recombinant
    Molecular Weight 95.5kd
    Purity ≥85% by SDS-PAGE
    Quantity 5μg
    Source Human
    Endotoxin Concentration <1 EU/μg
    Common Name Signal Transducer And Activator Of Transcription 6
    Gene Symbol STAT6
    Protein Tag His Tag
    Validated Application Assay Standard
    Protein Family Other Proteins
    Expression System Insect
    Product Type STAT6 Recombinant Human Protein
    For Use With (Application) Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications
    Format Lyophilized
    Gene ID (Entrez) 6778

    Suitable for use in protein studies such as protein structure analysis and protein-protein interactions

    • Formulation: 20mM Tris, 500mM NaCl, 20% Glycerol, 0.3mM DTT, pH 7.4

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit