Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

State of Pennsylvania BTEX Standard, High Level, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Manufacturer:  SPEX CertiPrepSupplier Diversity Partner PABTEXH

Catalog No. PA-BTEX-H


Add to cart

Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Benzene71-43-22000µg/mL
Ethylbenzene100-41-42000µg/mL
Isopropylbenzene98-82-82000µg/mL
Methyl-tert-butyl ether1634-04-42000µg/mL
m-Xylene108-38-32000µg/mL
Naphthalene91-20-32000µg/mL
o-Xylene95-47-62000µg/mL
p-Xylene106-42-32000µg/mL
Toluene108-88-32000µg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

BTEX Standard (High Level)
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
1mL
2mL amber prescored glass ampule
Methanol-P&T
GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Safety and Handling

Safety and Handling

DOTInformation : Nonhazardous

Recommended Storage : Ambient

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Product intended for laboratory use only