Learn More
Desco™ Statfree T2™ Dissipative Dual Layer Rubber Mat Top Kit
SDP
Dissipative Dual Layer Rubber Mat provides a worksurface that does not generate a static charge.
Supplier:  Desco™ 66040
Catalog No. 18-688-143
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Provides a worksurface that does not generate a static charge and will control the discharge rate from all conductors (including ESD susceptible items) that are placed on the surface.
  • Color (1 x 106 to < 1 x 109 ohms Rtt) Top Layer - Meets ANSI/ESD S20.20 worksurface required limit and recommendation of ANSI/ESD S4.1 for contact with ESD susceptible items
  • Electricals will not drift for the life of the mat
  • Black Side (4 Rtt) Bottom Layer - The dedicated conductive layer reduces the resistance to ground (Rtg) from the top of the surface to the groundable point ground and allows the mats to be used with ESD workstation monitors
  • High Strength Nitrile Rubber Compound - Designed to withstand abrasion, stretching, and tearing and may be used in soldering applications with flux and other chemicals
  • Ultraviolet (UV) Stabilizer Additives - Stabilizes color retention, ensures ESD performance over time and eliminates surface layer deterioration resulting in cracking when the product is flexed
  • Non-Humidity Dependence - Provides consistent electrical performance regardless of ambient humidity
  • Lead-free RoHS compliant
Specifications
Specifications
0.06 in.
24 in.
0.91 m
0.61 m
Blue
36 in.
Rubber