Learn More
GSC Go Science Crazy Static Charge Indicator Products for Science Education
Easily and effectively determine if a charge is positive or negative.
Manufacturer:  GSC Go Science Crazy ELSTCH02
Catalog No. S13927
Add to cart
Alert:
Not a toy. Product intended for ages 13 and up.
Description
Description
  • LED lights indicate if charge is positive or negative
  • 3 Friction Rods included - acrylic, PVC, and nylon
  • 5V power adapter included
Specifications
Specifications
Static Charge Indicator, Static Charge Indicator
3 friction rods, acrylic, PVC, nylon; Indicative LED lights; 5V power adapter
Education