Brady Corporation SORBENT PAD OIL 30X30IN 25/PK

Manufacturer: Brady Corporation SR3625

Catalog No. 17-111-139

  Check Availability
Add to Cart

    Specifications

    Product Type Static Resistant Heavy Pad
    Dimensions (L x W) 76.2 x 76.2cm (30 x 30 in.)
    For Use With (Application) Overspray leaks and drips, workstations, drums - liquid storage, outfalls and discharge ponds
    Capacity (English) Sorbing 16 gal./bale
    Color White
    Packaging Quantity 25/Bale

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit